Assay Details
| Gene Symbol: Prpf38a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172697.3 | NP_766285.2 | 3 - 4 | 631 | 108 | |
| GenBank mRNA | AK075925.1 | - | 3 - 4 | 623 | 108 | |
| AK169741.1 | - | 3 - 4 | 584 | 108 | ||
| BC024734.1 | - | 3 - 4 | 604 | 108 | ||
| BC025853.1 | - | 3 - 4 | 533 | 108 | ||
| BC139277.1 | - | 3 - 4 | 569 | 8862299 | 108 | |
| BC139305.1 | - | 3 - 4 | 567 | 8862327 | 108 | |
| CT010186.1 | - | 3 - 4 | 409 | 108 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23196699 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23196702 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23196700 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23196701 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462047 | BC059875.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462047 | BC139277.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462047 | BC139305.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462047 | AK169741.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462047 | BC024734.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462047 | BC059875.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462047 | BC139277.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462047 | BC139305.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462047 | AK169741.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462047 | BC024734.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 642422 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 55005 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 647939 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675892 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 134272 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368213 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76388 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 689550 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 476558 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 680254 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478901 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 80422 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 172752 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108466 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158856 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121716 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462047 | BC059875.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462047 | BC139277.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462047 | BC139305.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462047 | AK169741.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462047 | BC024734.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462047 | BC059875.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462047 | BC139277.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462047 | BC139305.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462047 | AK169741.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462047 | BC024734.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196702 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196699 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196694 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23196681 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05354112 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158856 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108466 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121716 | - | - |
| Whole Mouse Genome G4122A | A_51_P462047 | BC059875.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P462047 | BC139277.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P462047 | BC139305.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P462047 | AK169741.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P462047 | BC024734.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P462047 | BC059875.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P462047 | BC139277.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P462047 | BC139305.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P462047 | AK169741.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P462047 | BC024734.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P137145 | AK031401.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101612 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133880 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158856 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121716 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108466 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03900317 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03900309 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03362153 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03900318 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03900316 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03900315 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23196699 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23196702 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23196700 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23196701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23196586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03900309 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03900318 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03675263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196663 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196590 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196667 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196654 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03362152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03997520 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23196670 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10515015 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515021 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515019 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515024 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515018 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515023 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515025 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515020 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515017 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515016 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515014 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515022 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515013 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433565_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1444087_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443282_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10515012 | - | - |
| Mouse Genome 430 2.0 Array | 1443282_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1433565_at | - | - |
| Mouse Genome 430 2.0 Array | 1444087_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1433565_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1444087_at | - | - |
| Mouse Genome 430 2.0 Array | 1443282_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 230596_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 230596_at | - | - |
| Mouse Expression Set 430 | 1433565_PM_at | - | - |
| Mouse Expression Set 430 | 1433565_at | - | - |
| Mouse Expression Set 430 | 1443282_PM_at | - | - |
| Mouse Expression Set 430 | 1443282_at | - | - |
| Mouse Expression Set 430 | 1444087_at | - | - |
| Mouse Expression Set 430 | 1444087_PM_at | - | - |
| Murine Genome U74v2 Set | 104713_at | - | - |
| Murine Genome U74v2 Set | 104713_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433565_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433565_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433565_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433565_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444087_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444087_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443282_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443282_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 601307 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6940181 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6400347 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5860110 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006940181 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006400347 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005860110 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1243430 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2630704 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2599164 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2630704 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2599164 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1243430 | - | - |
| mouse-6 v1.1 (Illumina) | scl0002736.1_2-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00230596.1_33-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00230596.1_294-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5860110 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6940181 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6400347 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00230596.1_33-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00230596.1_294-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002736.1_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6940181 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6400347 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5860110 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2978570 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2630704 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2599164 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2978574 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2978570 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2630704 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2599164 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2978574 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00230596.1_33-S | - | - |

Prpf38a
Genomic Map