Assay Details
| Gene Symbol: Sel1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172710.3 | NP_766298.2 | 7 - 8 | 1453 | 64 | |
| GenBank mRNA | AK030487.1 | - | 7 - 8 | 1318 | 64 | |
| AK037220.1 | - | 7 - 8 | 1317 | 64 | ||
| AK122362.1 | - | 7 - 8 | 1453 | 64 | ||
| BC094607.1 | - | 6 - 7 | 998 | 6837048 | 64 | |
| BC117915.1 | - | 6 - 7 | 954 | 40097358 | 64 | |
| BC117916.1 | - | 6 - 7 | 956 | 40097359 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sel1l3
Genomic Map