Assay Details
| Gene Symbol: Sel1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172710.3 | NP_766298.2 | 15 - 16 | 2617 | 65 | |
| GenBank mRNA | AK030487.1 | - | 15 - 16 | 2482 | 65 | |
| AK122362.1 | - | 15 - 16 | 2617 | 65 | ||
| BC026655.1 | - | 3 - 4 | 340 | 65 | ||
| BC094607.1 | - | 14 - 15 | 2162 | 6837048 | 65 | |
| BC117915.1 | - | 14 - 15 | 2118 | 40097358 | 65 | |
| BC117916.1 | - | 14 - 15 | 2120 | 40097359 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sel1l3
Genomic Map