Assay Details
| Gene Symbol: Guf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172711.2 | NP_766299.1 | 2 - 3 | 330 | 117 | |
| GenBank mRNA | AK032597.1 | - | 1 - 2 | 1338 | 117 | |
| AK047411.1 | - | 2 - 3 | 199 | 117 | ||
| AK053420.1 | - | 2 - 3 | 304 | 117 | ||
| AK084627.1 | - | 2 - 3 | 311 | 117 | ||
| AK160956.1 | - | 2 - 3 | 349 | 117 | ||
| AK165012.1 | - | 2 - 3 | 319 | 117 | ||
| BC057962.1 | - | 2 - 3 | 284 | 5353891 | 117 | |
| BY743247.1 | - | 2 - 3 | 331 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Guf1
Genomic Map