Assay Details
Gene Symbol: Guf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172711.2 | NP_766299.1 | 3 - 4 | 480 | 97 | |
GenBank mRNA | AK032597.1 | - | 2 - 3 | 1488 | 97 | |
AK047411.1 | - | 3 - 4 | 349 | 97 | ||
AK053420.1 | - | 3 - 4 | 454 | 97 | ||
AK084627.1 | - | 3 - 4 | 461 | 97 | ||
AK140534.1 | - | 3 - 4 | 439 | 97 | ||
AK160956.1 | - | 3 - 4 | 499 | 97 | ||
AK165012.1 | - | 3 - 4 | 469 | 97 | ||
BC057962.1 | - | 3 - 4 | 434 | 5353891 | 97 | |
BY743247.1 | - | 3 - 4 | 481 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
