Assay Details
Gene Symbol: Guf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172711.2 | NP_766299.1 | 4 - 5 | 561 | 69 | |
GenBank mRNA | AK032597.1 | - | 3 - 4 | 1569 | 69 | |
AK047411.1 | - | 4 - 5 | 430 | 69 | ||
AK053420.1 | - | 4 - 5 | 535 | 69 | ||
AK084627.1 | - | 4 - 5 | 542 | 69 | ||
AK140534.1 | - | 4 - 5 | 520 | 69 | ||
AK160956.1 | - | 4 - 5 | 580 | 69 | ||
AK165012.1 | - | 4 - 5 | 550 | 69 | ||
BC057962.1 | - | 4 - 5 | 515 | 5353891 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
