Assay Details
| Gene Symbol: Guf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172711.2 | NP_766299.1 | 5 - 6 | 651 | 76 | |
| GenBank mRNA | AK032597.1 | - | 4 - 5 | 1659 | 76 | |
| AK047411.1 | - | 5 - 6 | 520 | 76 | ||
| AK053420.1 | - | 5 - 6 | 625 | 76 | ||
| AK084627.1 | - | 5 - 6 | 632 | 76 | ||
| AK140534.1 | - | 5 - 6 | 610 | 76 | ||
| AK160956.1 | - | 5 - 6 | 670 | 76 | ||
| AK165012.1 | - | 5 - 6 | 640 | 76 | ||
| BC057962.1 | - | 5 - 6 | 605 | 5353891 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Guf1
Genomic Map