Assay Details
| Gene Symbol: Guf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172711.2 | NP_766299.1 | 9 - 10 | 1136 | 84 | |
| GenBank mRNA | AK053420.1 | - | 9 - 10 | 1110 | 84 | |
| AK084627.1 | - | 9 - 10 | 1117 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Guf1
Genomic Map