Assay Details
| Gene Symbol: Adap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172723.4 | NP_766311.2 | 5 - 6 | 599 | 67 | |
| GenBank mRNA | AK076754.1 | - | 4 - 5 | 585 | 67 | |
| AK076768.1 | - | 5 - 6 | 356 | 67 | ||
| BC056934.1 | - | 4 - 5 | 325 | 67 | ||
| BC079634.1 | - | 4 - 5 | 393 | 67 | ||
| BC144874.1 | - | 4 - 5 | 344 | 67 | ||
More Information
Set Membership: |
|

Adap1
Genomic Map