Assay Details
Gene Symbol: Brat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276287.1 | NP_001263216.1 | 5 - 6 | 981 | 61 | |
NM_181066.3 | NP_851411.1 | 5 - 6 | 883 | 61 | ||
NR_074085.1 | 5 - 6 | 883 | 61 | |||
GenBank mRNA | AK049321.1 | - | 5 - 6 | 872 | 61 | |
AK084987.1 | - | 5 - 6 | 872 | 61 | ||
AK085091.1 | - | 5 - 6 | 872 | 61 | ||
AK154448.1 | - | 5 - 6 | 854 | 61 | ||
AK170269.1 | - | 5 - 6 | 877 | 61 | ||
BC068301.1 | - | 5 - 6 | 947 | 30061848 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
