Assay Details
Gene Symbol: Brat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276287.1 | NP_001263216.1 | 7 - 8 | 1195 | 75 | |
NM_181066.3 | NP_851411.1 | 7 - 8 | 1097 | 75 | ||
GenBank mRNA | AK043212.1 | - | 7 - 8 | 1084 | 75 | |
AK084987.1 | - | 7 - 8 | 1086 | 75 | ||
AK085091.1 | - | 7 - 8 | 1086 | 75 | ||
AK154448.1 | - | 7 - 8 | 1068 | 75 | ||
AK170269.1 | - | 7 - 8 | 1091 | 75 | ||
BC068301.1 | - | 7 - 8 | 1161 | 30061848 | 75 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
