Assay Details
| Gene Symbol: Mtmr10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172742.2 | NP_766330.2 | 13 - 14 | 1477 | 66 | |
| GenBank mRNA | AK028584.1 | - | 1 - 2 | 102 | 66 | |
| BC041672.1 | - | 13 - 14 | 1449 | 66 | ||
| BC055074.1 | - | 11 - 12 | 1141 | 30052208 | 66 | |
| BC058196.1 | - | 6 - 7 | 579 | 66 | ||
| CB600359.1 | - | 1 - 2 | 148 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtmr10
Genomic Map