Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900022539.mm.1 |
NM_001276452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022539.mm.1 |
NM_172773.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022539.mm.1 |
TC0900002728.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900022540.mm.1 |
NM_172773.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022540.mm.1 |
TC0900002728.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900022541.mm.1 |
NM_001276452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022541.mm.1 |
NM_172773.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022541.mm.1 |
TC0900002728.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900022542.mm.1 |
NM_001276452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022542.mm.1 |
NM_172773.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022542.mm.1 |
TC0900002728.mm.1 |
MTA TC |