Assay Details
| Gene Symbol: Atrip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172774.3 | NP_766362.2 | 6 - 7 | 1050 | 75 | |
| GenBank mRNA | AK031257.1 | - | 6 - 7 | 1040 | 75 | |
| AK171423.1 | - | 6 - 7 | 1005 | 75 | ||
| BC078458.1 | - | 6 - 7 | 997 | 6832436 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atrip
Genomic Map