Assay Details
Gene Symbol: Ddx26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172779.4 | NP_766367.1 | 1 - 2 | 483 | 79 | |
GenBank mRNA | AK031945.1 | - | 1 - 2 | 475 | 79 | |
AK083942.1 | - | 1 - 2 | 445 | 79 | ||
AK160627.1 | - | 1 - 2 | 443 | 79 | ||
BC051161.1 | - | 1 - 2 | 465 | 6466142 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
