Assay Details
Gene Symbol: Ddx26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172779.4 | NP_766367.1 | 14 - 15 | 2373 | 81 | |
GenBank mRNA | AK083942.1 | - | 14 - 15 | 2335 | 81 | |
AK144026.1 | - | 4 - 5 | 2218 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
