Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100030401.mm.1 |
NM_001168521.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030401.mm.1 |
NM_001168521.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030401.mm.1 |
TC1100003297.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030399.mm.1 |
NM_001168521.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030399.mm.1 |
NM_001168521.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030399.mm.1 |
NM_172795.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030399.mm.1 |
NM_172795.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030399.mm.1 |
TC1100003297.mm.1 |
MTA TC |