Assay Details
| Gene Symbol: Otop2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172801.2 | NP_766389.2 | 3 - 4 | 639 | 72 | |
| GenBank mRNA | AB087504.1 | - | 2 - 3 | 524 | 72 | |
| AK028866.1 | - | 3 - 4 | 640 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Otop2
Genomic Map