Assay Details
Gene Symbol: Antxrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172808.2 | NP_766396.2 | 8 - 9 | 933 | 104 | |
GenBank mRNA | AK077206.1 | - | 8 - 9 | 934 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
