Assay Details
Gene Symbol: Lnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172827.3 | NP_766415.1 | 4 - 5 | 1209 | 96 | |
GenBank mRNA | AK029094.1 | - | 4 - 5 | 1210 | 96 | |
AK036967.1 | - | 4 - 5 | 1210 | 96 | ||
AK161484.1 | - | 436 | 96 | |||
AK162332.1 | - | 1086 | 96 | |||
BC066791.1 | - | 5 - 6 | 1533 | 30054751 | 96 | |
BC120925.1 | - | 4 - 5 | 1166 | 40098425 | 96 | |
BC120926.1 | - | 4 - 5 | 1166 | 40098428 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
