Assay Details
Gene Symbol: Lnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172827.3 | NP_766415.1 | 8 - 9 | 1728 | 137 | |
GenBank mRNA | AK029094.1 | - | 8 - 9 | 1729 | 137 | |
AK036967.1 | - | 8 - 9 | 1729 | 137 | ||
AK161484.1 | - | 955 | 137 | |||
BC066791.1 | - | 9 - 10 | 2052 | 30054751 | 137 | |
BC120925.1 | - | 8 - 9 | 1685 | 40098425 | 137 | |
BC120926.1 | - | 8 - 9 | 1685 | 40098428 | 137 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
