Assay Details
| Gene Symbol: Peli3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172835.3 | NP_766423.3 | 5 - 6 | 741 | 64 | |
| GenBank mRNA | AK044418.1 | - | 5 - 6 | 688 | 64 | |
| AK135575.1 | - | 4 - 5 | 588 | 64 | ||
| BC028931.1 | - | 5 - 6 | 699 | 5037308 | 64 | |
More Information
Set Membership: |
|

Peli3
Genomic Map