Assay Details
| Gene Symbol: Lipk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205349.1 | NP_001192278.1 | 3 - 4 | 377 | 62 | |
| NM_172837.4 | NP_766425.1 | 3 - 4 | 362 | 62 | ||
| GenBank mRNA | AK037106.1 | - | 3 - 4 | 327 | 62 | |
| AK162706.1 | - | 3 - 4 | 284 | 62 | ||
| BC037229.1 | - | 4 - 5 | 329 | 62 | ||
| BC055815.1 | - | 3 - 4 | 330 | 4011597 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lipk
Genomic Map