Assay Details
Gene Symbol: Dnah14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK038967.1 | - | 7 - 8 | 1014 | 71 | |
AK043618.1 | - | 8 - 9 | 1021 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P111655 | AK038967.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037463 | AK043618.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 572898 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206346 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110021 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174085 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P111655 | AK038967.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037463 | AK043618.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04378246 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174085 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110021 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04378260 | - | - |
Whole Mouse Genome G4122A | A_52_P317273 | AK043618.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P317273 | AK038967.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P216696 | AK038967.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111655 | AK038967.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134555 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110021 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883416 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883256 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883225 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883340 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883208 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883201 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883350 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883433 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883262 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20883341 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883246 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883352 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00727010 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883358 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883235 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00125268 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00916008 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883282 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883447 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883292 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883350 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00727004 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883240 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00685299 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883413 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00504687 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883252 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883223 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883417 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883217 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883441 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883210 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00836003 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883226 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00445642 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00504676 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883409 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883200 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883272 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00504671 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883356 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883251 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883268 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00714153 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00916006 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883199 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883406 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883233 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883205 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00843869 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883277 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883220 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883289 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00727007 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883448 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883249 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883284 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883229 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883360 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00631908 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883213 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883238 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883411 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00000370 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883275 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883405 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883351 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00326729 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883353 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883236 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883414 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883206 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00631907 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883364 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883452 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883287 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883221 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00727006 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883258 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883291 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883256 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883225 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883445 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00714156 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883257 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883410 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883241 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883201 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883362 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883211 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00749715 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883208 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00125269 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883237 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883215 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883203 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883259 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00125265 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883219 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883439 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883262 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20883418 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 609071 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 3190215 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003190215 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2714837 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2714837 | - | - |
mouse-6 v1.1 (Illumina) | scl17121.12.1_29-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3190215 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl17121.12.1_29-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3190215 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1218082 | - | - |
