Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200007795.mm.1 |
NM_172856.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007795.mm.1 |
TC0200000994.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007797.mm.1 |
NM_172856.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007797.mm.1 |
TC0200000994.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007793.mm.1 |
TC0200000994.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007799.mm.1 |
NM_172856.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007799.mm.1 |
TC0200000994.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007794.mm.1 |
NM_172856.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007794.mm.1 |
TC0200000994.mm.1 |
MTA TC |