Assay Details
| Gene Symbol: Exd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172857.2 | NP_766445.1 | 4 - 5 | 967 | 136 | |
| GenBank mRNA | AK030132.1 | - | 4 - 5 | 969 | 136 | |
| BC092241.1 | - | 2 - 3 | 221 | 30670567 | 136 | |
| BC144750.1 | - | 3 - 4 | 350 | 9053298 | 136 | |
| BC150662.1 | - | 4 - 5 | 444 | 9087573 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exd1
Genomic Map