Assay Details
Gene Symbol: Pak7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172858.2 | NP_766446.2 | 7 - 8 | 2226 | 78 | |
GenBank mRNA | AK032593.1 | - | 7 - 8 | 2199 | 78 | |
AK079080.1 | - | 8 - 9 | 2319 | 78 | ||
AK163530.1 | - | 7 - 8 | 2034 | 78 | ||
AY487425.1 | - | 5 - 6 | 1746 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
