Assay Details
| Gene Symbol: Slc13a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172892.3 | NP_766480.1 | 8 - 9 | 1565 | 87 | |
| GenBank mRNA | AK036354.1 | - | 8 - 9 | 1567 | 87 | |
| AK038937.1 | - | 8 - 9 | 1570 | 87 | ||
| BC089161.1 | - | 8 - 9 | 1554 | 30440173 | 87 | |
More Information
Set Membership: |
|

Slc13a4
Genomic Map