Assay Details
Gene Symbol: D8Ertd82e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172911.3 | NP_766499.2 | 3 - 4 | 2374 | 108 | |
GenBank mRNA | AK036672.1 | - | 3 - 4 | 2342 | 108 | |
AK220205.1 | - | 1 - 2 | 1522 | 108 | ||
BC089024.1 | - | 2 - 3 | 1832 | 30615630 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
