Assay Details
| Gene Symbol: Amdhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172935.4 | NP_766523.2 | 3 - 4 | 429 | 137 | |
| GenBank mRNA | AK077584.1 | - | 3 - 4 | 412 | 137 | |
| AK090009.1 | - | 3 - 4 | 431 | 137 | ||
| AK154209.1 | - | 3 - 4 | 429 | 137 | ||
| AK159860.1 | - | 3 - 4 | 426 | 137 | ||
| AK168432.1 | - | 3 - 4 | 387 | 137 | ||
| BC037005.1 | - | 3 - 4 | 465 | 5101453 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|

Amdhd2
Genomic Map