Assay Details
| Gene Symbol: Amdhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172935.4 | NP_766523.2 | 7 - 8 | 934 | 67 | |
| GenBank mRNA | AK077584.1 | - | 7 - 8 | 917 | 67 | |
| AK090009.1 | - | 7 - 8 | 936 | 67 | ||
| AK154209.1 | - | 7 - 8 | 935 | 67 | ||
| AK168432.1 | - | 7 - 8 | 892 | 67 | ||
| AK189145.1 | - | 2 - 3 | 217 | 67 | ||
| AK200617.1 | - | 2 - 3 | 253 | 67 | ||
| BC037005.1 | - | 7 - 8 | 970 | 5101453 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Amdhd2
Genomic Map