Assay Details
| Gene Symbol: Mtmr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172958.3 | NP_766546.1 | 3 - 4 | 392 | 73 | |
| GenBank mRNA | AK030139.1 | - | 3 - 4 | 459 | 73 | |
| AK050099.1 | - | 3 - 4 | 385 | 73 | ||
| AK050236.1 | - | 3 - 4 | 383 | 73 | ||
| AK122539.1 | - | 3 - 4 | 450 | 73 | ||
| BC042573.1 | - | 3 - 4 | 381 | 73 | ||
| BC054352.1 | - | 3 - 4 | 392 | 3661020 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtmr12
Genomic Map