Assay Details
Gene Symbol: Map1s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173013.3 | NP_766601.2 | 5 - 6 | 2568 | 107 | |
GenBank mRNA | AK032300.1 | - | 5 - 6 | 2556 | 107 | |
AK042159.1 | - | 5 - 6 | 2565 | 107 | ||
AK082854.1 | - | 5 - 6 | 2563 | 107 | ||
AK162124.1 | - | 1 - 2 | 1276 | 107 | ||
BC052828.1 | - | 1 - 2 | 1415 | 107 | ||
BC079598.1 | - | 3 - 4 | 3929 | 107 | ||
BC150729.1 | - | 5 - 6 | 2610 | 9087640 | 107 | |
DQ387862.1 | - | 5 - 6 | 2556 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
