Assay Details
Gene Symbol: Pfkfb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173019.5 | NP_766607.3 | 11 - 12 | 1332 | 81 | |
GenBank mRNA | AK048999.1 | - | 3 - 4 | 446 | 81 | |
AK155172.1 | - | 11 - 12 | 1270 | 81 | ||
AY646092.2 | - | 11 - 12 | 1218 | 81 | ||
AY707862.1 | - | 11 - 12 | 1235 | 81 | ||
AY756065.1 | - | 11 - 12 | 1325 | 81 | ||
AY756066.1 | - | 11 - 12 | 1325 | 81 | ||
AY756067.1 | - | 11 - 12 | 1196 | 81 | ||
BC057594.1 | - | 11 - 12 | 1284 | 6810667 | 81 | |
DQ350638.1 | - | 12 - 13 | 1304 | 81 | ||
EU571215.1 | - | 11 - 12 | 1316 | 81 | ||
EU571216.1 | - | 13 - 14 | 1491 | 81 | ||
EU571217.1 | - | 11 - 12 | 1459 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
