Assay Details
| Gene Symbol: P3h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173379.2 | NP_775555.1 | 9 - 10 | 1536 | 71 | |
| GenBank mRNA | AJ430350.1 | - | 9 - 10 | 1537 | 71 | |
| AK076434.1 | - | 554 | 71 | |||
More Information
Set Membership: |
|

P3h2
Genomic Map