Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300002735.mm.1 |
NM_173382.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002735.mm.1 |
NM_173382.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002735.mm.1 |
TC0300000396.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300002736.mm.1 |
NM_173382.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002736.mm.1 |
NM_173382.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002736.mm.1 |
TC0300000396.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300002737.mm.1 |
NM_173382.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002737.mm.1 |
NM_173382.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002737.mm.1 |
TC0300000396.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300002738.mm.1 |
NM_173382.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002738.mm.1 |
NM_173382.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002738.mm.1 |
TC0300000396.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300002740.mm.1 |
NM_173382.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002740.mm.1 |
NM_173382.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002740.mm.1 |
TC0300000396.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300002739.mm.1 |
NM_173382.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002739.mm.1 |
NM_173382.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300002739.mm.1 |
TC0300000396.mm.1 |
MTA TC |