Assay Details
| Gene Symbol: Klhdc7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173427.2 | NP_775603.2 | 1 | 1979 | 117 | |
| GenBank mRNA | AK045732.1 | - | 1 | 1982 | 117 | |
| AK050048.1 | - | 1 | 1566 | 117 | ||
| AK052747.1 | - | 1 | 1549 | 117 | ||
| BC043462.1 | - | 1 | 518 | 117 | ||
| BC043672.1 | - | 1 | 518 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23355138 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23355140 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23355139 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23355141 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963374 | AW124675.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963374 | NM_173427.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 311223 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 890880 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884089 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 286889 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 64150 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 46355 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116096 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963374 | AW124675.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963374 | NM_173427.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23355147 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116096 | - | - |
| Whole Mouse Genome G4122A | A_52_P628369 | BC043672.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P628369 | AK052747.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P628369 | BC043462.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P628369 | NM_173427.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P628369 | AK045732.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P424990 | BC043672.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P424990 | BC043462.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P424990 | NM_173427.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P424990 | AK045732.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129133 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116096 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03900392 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03238828 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03900433 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03900442 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03900424 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03900421 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03900428 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23355139 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23355141 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23355138 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23355140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23355094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03325106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03224481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355196 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03238100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03900436 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03862473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03238089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03267358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355187 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03900400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03238829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03900394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03900439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355102 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03725958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03900412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03763831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03900426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03763835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03763837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03900418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355172 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03900441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03763838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03267363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23355199 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10517729 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517730 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517728 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456409_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10517727 | - | - |
| Mouse Genome 430 2.0 Array | 1456409_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456409_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 242721_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 242721_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400031780.mm.1 | TC0400003855.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400031780.mm.1 | NM_173427.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031780.mm.1 | NM_173427.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031781.mm.1 | TC0400003855.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400031781.mm.1 | NM_173427.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031781.mm.1 | NM_173427.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031781.mm.1 | NR_035519.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031782.mm.1 | TC0400003855.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400031782.mm.1 | NM_173427.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031782.mm.1 | NM_173427.2 | Refseq |
| Mouse Expression Set 430 | 1456409_at | - | - |
| Mouse Expression Set 430 | 1456409_PM_at | - | - |
| Murine Genome U74v2 Set | 139138_at | - | - |
| Murine Genome U74v2 Set | 139138_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456409_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456409_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 854101 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106590059 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106590059 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 430086 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000430086 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1234069 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234069 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254436 | - | - |
| mouse-6 v1.1 (Illumina) | scl23616.1_437-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00242721.2_229-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106590059 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 430086 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00242721.2_229-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106590059 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 430086 | - | - |

Klhdc7a
Genomic Map