Assay Details
| Gene Symbol: Rccd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173445.4 | NP_775621.2 | 1 - 2 | 704 | 82 | |
| GenBank mRNA | AK030872.1 | - | 1 - 2 | 675 | 82 | |
| AK084219.1 | - | 1 - 2 | 379 | 82 | ||
| AK088482.1 | - | 1 - 2 | 705 | 82 | ||
| AK088662.1 | - | 1 - 2 | 345 | 82 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK030872.1,AK084219.1,AK088662.1,NM_173445.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25266018 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25266017 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25266015 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016466 | BC069844.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016466 | AK088662.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016466 | AK084219.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016466 | NM_173445.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016466 | AK030872.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 268147 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 272695 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 954300 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 84799 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 662225 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 44687 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P176936 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016466 | BC069844.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016466 | AK088662.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016466 | AK084219.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016466 | NM_173445.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016466 | AK030872.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25266017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25266010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25266023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25266012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25266020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25266016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25266006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25266014 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P176936 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06256106 | - | - |
| Whole Mouse Genome G4122A | A_52_P301035 | BC069844.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P301035 | AK088482.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P301035 | AK088662.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P301035 | AK047578.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P301035 | AK084219.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P27635 | AK047578.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134659 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P176936 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06095199 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06095202 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06095213 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25266017 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25266015 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25266018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265906 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06095207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06095213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06202680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06202706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06202694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06043856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06288710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06202689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06202690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06095202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266012 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10564887 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564885 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564878 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564886 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564883 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564880 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564884 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564881 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564882 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564879 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10564877 | - | - |
| Mouse Genome 430 2.0 Array | 1456411_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456411_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269955_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269955_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700030355.mm.1 | NM_173445.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700030355.mm.1 | NM_173445.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700030355.mm.1 | TC0700003590.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700030347.mm.1 | NM_173445.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700030347.mm.1 | NM_173445.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700030347.mm.1 | TC0700003590.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700030351.mm.1 | NM_173445.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700030351.mm.1 | NM_173445.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700030351.mm.1 | TC0700003590.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700030356.mm.1 | NM_173445.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700030356.mm.1 | NM_173445.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700030356.mm.1 | TC0700003590.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700030350.mm.1 | NM_173445.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700030350.mm.1 | NM_173445.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700030350.mm.1 | TC0700003590.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1456411_at | - | - |
| Mouse Expression Set 430 | 1456411_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456411_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456411_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 431038 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 610408 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2360044 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 610408 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000610408 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002360044 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1213882 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2691271 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1213882 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2691271 | - | - |
| mouse-6 v1.1 (Illumina) | scl00269955.1_25-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0269955.3_29-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 610408 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2360044 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0269955.3_29-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00269955.1_25-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 610408 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2360044 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2838629 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2691271 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1213882 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2838629 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2691271 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1213882 | - | - |

Rccd1
Genomic Map