Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900024293.mm.1 |
NM_001168296.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024293.mm.1 |
NM_001168296.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024293.mm.1 |
NM_173447.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024293.mm.1 |
NM_173447.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024293.mm.1 |
TC0900002956.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024294.mm.1 |
NM_001168296.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024294.mm.1 |
NM_001168296.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024294.mm.1 |
NM_173447.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024294.mm.1 |
NM_173447.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024294.mm.1 |
TC0900002956.mm.1 |
MTA TC |