Assay Details
| Gene Symbol: Maoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173740.3 | NP_776101.3 | 11 - 12 | 1307 | 67 | |
| GenBank mRNA | AK078642.1 | - | 4 - 5 | 294 | 67 | |
| AK146612.1 | - | 11 - 12 | 1308 | 67 | ||
| AK155052.1 | - | 11 - 12 | 1170 | 67 | ||
| AK164457.1 | - | 11 - 12 | 1144 | 67 | ||
| BC029100.1 | - | 11 - 12 | 1177 | 3481702 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Maoa
Genomic Map