Assay Details
| Gene Symbol: Cdh22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174988.3 | NP_778153.2 | 9 - 10 | 2030 | 68 | |
| GenBank mRNA | AB019618.1 | - | 9 - 10 | 2020 | 68 | |
| AK140248.1 | - | 9 - 10 | 2148 | 68 | ||
| BC112407.1 | - | 8 - 9 | 1663 | 40060660 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdh22
Genomic Map