Assay Details
| Gene Symbol: Atp5g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301721.1 | NP_001288650.1 | 4 - 5 | 461 | 66 | |
| NM_175015.3 | NP_778180.1 | 4 - 5 | 451 | 66 | ||
| GenBank mRNA | AI851824.1 | - | 1 - 2 | 66 | ||
| AK077930.1 | - | 4 - 5 | 420 | 66 | ||
| AK087955.1 | - | 4 - 5 | 428 | 66 | ||
| BC116218.2 | - | 3 - 4 | 381 | 40091232 | 66 | |
| CF104340.1 | - | 4 - 5 | 447 | 66 | ||
| CX208968.1 | - | 4 - 5 | 418 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21334262 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21334260 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21334259 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21334257 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21334258 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21334261 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046703 | NM_175015.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046703 | AK077930.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046703 | AK087955.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 746623 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 487443 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84286 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P176916 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046703 | NM_175015.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046703 | AK077930.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046703 | AK087955.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P294849 | NM_175015.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P294849 | AK077930.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P294849 | AK087955.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21334269 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P176916 | - | - |
| Whole Mouse Genome G4122A | A_51_P294849 | NM_175015.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P294849 | AK077930.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P294849 | AK087955.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115266 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P176916 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01593492 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01955785 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02088627 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02081668 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21334257 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21334261 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21334262 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21334258 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21334260 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21334259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21334223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334233 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334296 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334236 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02063125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02088632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01279134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02088627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01654441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02088639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334225 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02088633 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334241 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01875231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02088647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334256 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334280 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02233235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21334281 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10483758 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10483759 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10483760 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10483757 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454661_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10483756 | - | - |
| Mouse Genome 430 2.0 Array | 1454661_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454661_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 228033_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 228033_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200032241.mm.1 | NM_175015.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200032241.mm.1 | TC0200003765.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200032242.mm.1 | NM_175015.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200032242.mm.1 | TC0200003765.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1454661_PM_at | - | - |
| Mouse Expression Set 430 | 1454661_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454661_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454661_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1454661_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1454661_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4230551 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4230551 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004230551 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2692124 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2692124 | - | - |
| mouse-6 v1.1 (Illumina) | scl19131.5.1_9-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4230551 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl19131.5.1_9-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4230551 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2692124 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2692124 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl19131.5.1_9-S | - | - |

Atp5g3
Genomic Map