Assay Details
Gene Symbol: Prickle3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290624.1 | NP_001277553.1 | 3 - 4 | 390 | 85 | |
GenBank mRNA | AK037217.1 | - | 3 - 4 | 353 | 85 | |
AK083704.1 | - | 2 - 3 | 357 | 85 | ||
BC023671.1 | - | 3 - 4 | 386 | 85 | ||
BC049258.1 | - | 3 - 4 | 332 | 5250488 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P133792 | BC032173.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P133792 | BC049258.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P133792 | AK083704.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P133792 | NM_175097.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P133792 | AK037217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P133792 | BC032173.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P133792 | BC049258.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P133792 | AK083704.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P133792 | NM_175097.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P133792 | AK037217.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 861504 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 778424 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 693560 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 963762 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 659001 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 933031 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 218089 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 467725 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107040 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136421 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P133792 | BC032173.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P133792 | BC049258.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P133792 | AK083704.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P133792 | NM_175097.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P133792 | AK037217.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P133792 | BC032173.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P133792 | BC049258.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P133792 | AK083704.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P133792 | NM_175097.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P133792 | AK037217.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239493 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239514 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239494 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239511 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239523 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239518 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239508 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239509 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239521 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239520 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239499 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239522 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239501 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239519 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239505 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239517 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239500 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239516 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239512 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239492 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239504 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239497 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239496 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09226452 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107040 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136421 | - | - |
Whole Mouse Genome G4122A | A_51_P133792 | BC032173.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P133792 | BC049258.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P133792 | AK083704.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P133792 | NM_175097.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P133792 | AK037217.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P133792 | BC032173.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P133792 | BC049258.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P133792 | AK083704.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P133792 | NM_175097.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P133792 | AK037217.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P1048 | AK079845.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131046 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107040 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136421 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14878200 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14878209 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14878182 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14878207 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14878181 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14878205 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14878201 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14878178 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14878208 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14878206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239520 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239500 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239564 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239632 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239556 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239544 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239547 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239587 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239601 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239582 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239574 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239569 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239561 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239493 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239505 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239585 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239559 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239558 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239603 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239511 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239622 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239501 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14963601 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239514 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14643972 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14963605 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14963611 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239512 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239508 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239504 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239536 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239562 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239569 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239559 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14714456 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239496 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239533 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239522 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239558 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239517 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239521 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14641340 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14746048 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239509 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14763943 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239549 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239523 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14878177 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239511 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239543 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239565 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239544 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14963620 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239547 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239531 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239548 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239530 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239542 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239497 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14864034 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239541 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239568 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239537 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239499 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239550 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14878176 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239500 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239546 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239525 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239561 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14643977 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32239520 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10598374 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10598371 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10411360 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10598377 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10598375 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10598376 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10598373 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10598378 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10603348 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10598379 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10603347 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10598380 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10598372 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1439296_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10598369 | - | - |
Mouse Genome 430 2.0 Array | 1439296_at | - | - |
Mouse Genome 430 2.0 Array | 1439296_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0X00000321.mm.1 | TC0X00000061.mm.1 | MTA TC |
Mouse Expression Set 430 | 1439296_at | - | - |
Mouse Expression Set 430 | 1439296_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1439296_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1439296_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1439296_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1439296_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 928427 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4210072 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4210072 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004210072 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2739398 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2602858 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2602858 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2739398 | - | - |
mouse-6 v1.1 (Illumina) | scl55078.9.1_18-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4210072 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl55078.9.1_18-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4210072 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2739398 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2739398 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl55078.9.1_18-S | - | - |
