Assay Details
Gene Symbol: Rae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175112.5 | NP_780321.1 | 9 - 10 | 1019 | 70 | |
GenBank mRNA | AK014325.1 | - | 9 - 10 | 975 | 70 | |
AK017746.1 | - | 9 - 10 | 940 | 70 | ||
AK035278.1 | - | 9 - 10 | 965 | 70 | ||
AK079383.1 | - | 9 - 10 | 1000 | 70 | ||
AK146058.1 | - | 9 - 10 | 984 | 70 | ||
AK156681.1 | - | 9 - 10 | 984 | 70 | ||
BC059051.1 | - | 9 - 10 | 941 | 6818771 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
