Assay Details
| Gene Symbol: Gpr61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK032627.1 | - | 2 | 1015 | 64 | |
| AK133702.1 | - | 2 | 1014 | 64 | ||
| BC116333.2 | - | 1 | 671 | 40096913 | 64 | |
| BC116334.2 | - | 1 | 671 | 40096914 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148624 | BC116333.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148624 | BC116334.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148624 | AK032627.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148624 | AK133702.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148624 | NM_175470.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2185618 | AK032627.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2185618 | NM_175470.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 233807 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877417 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 629243 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 525074 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 753346 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 83930 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152576 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148624 | BC116333.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148624 | BC116334.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148624 | AK032627.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148624 | AK133702.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148624 | NM_175470.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2185618 | AK032627.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2185618 | NM_175470.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P149094 | BC116333.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P149094 | BC116334.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P149094 | AK032627.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P149094 | NM_175470.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410943 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410922 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22410888 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00722819 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152576 | - | - |
| Whole Mouse Genome G4122A | A_51_P149094 | BC116333.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P149094 | BC116334.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P149094 | AK032627.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P149094 | NM_175470.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124115 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152576 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02879251 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03111531 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03111529 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02879248 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02879260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22410832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02879251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410931 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02704035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03121073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410944 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02704034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02906948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03121076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03121070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02879250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02879244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02704016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03121074 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02704033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02704014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02704038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410941 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03184434 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22410912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03010734 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10501279 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501280 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501278 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501281 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436972_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10501277 | - | - |
| Mouse Genome 430 2.0 Array | 1436972_at | - | - |
| Mouse Genome 430 2.0 Array | 1436972_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300021425.mm.1 | TC0300002732.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021424.mm.1 | TC0300002732.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021427.mm.1 | TC0300002732.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021426.mm.1 | TC0300002732.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1436972_at | - | - |
| Mouse Expression Set 430 | 1436972_PM_at | - | - |
| Murine Genome U74v2 Set | 169242_PM_at | - | - |
| Murine Genome U74v2 Set | 169242_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436972_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436972_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 664128 | - | - |
| ABI Mouse Genome Survey Microarray | 744742 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6980035 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6980035 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006980035 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1254930 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254930 | - | - |
| mouse-6 v1.1 (Illumina) | scl21658.2_367-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6980035 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21658.2_367-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6980035 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1254930 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254930 | - | - |

Gpr61
Genomic Map