Assay Details
| Gene Symbol: Hopx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175606.3 | NP_783199.1 | 1 - 2 | 272 | 82 | |
| GenBank mRNA | AF492703.1 | - | 1 - 2 | 272 | 82 | |
| AF534182.1 | - | 1 - 2 | 317 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF492703.1, AF534182.1, NM_175606.3 transcripts.
Set Membership: |
|

Hopx
Genomic Map