Assay Details
| Gene Symbol: Hist2h2be | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178214.3 | NP_835586.2 | 1 | 455 | 70 | |
| GenBank mRNA | AK078533.1 | - | 1 | 456 | 70 | |
| AK090189.1 | - | 1 | 458 | 70 | ||
| AK162494.1 | - | 1 | 459 | 70 | ||
| AK167546.1 | - | 1 | 458 | 70 | ||
| AK168767.1 | - | 1 | 455 | 70 | ||
| AK170749.1 | - | 1 | 458 | 70 | ||
| BC047137.1 | - | 1 | 435 | 6311571 | 70 | |
| BC058562.1 | - | 1 | 457 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22348231 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22348232 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2177488 | BC047137.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2177488 | BC058562.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2177488 | AK168767.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2177488 | AK170749.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2177488 | AK162494.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 532777 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146591 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2177488 | BC047137.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2177488 | BC058562.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2177488 | AK168767.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2177488 | AK170749.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2177488 | AK162494.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430071 | BC047137.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430071 | BC058562.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430071 | AK162494.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430071 | AK167546.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P430071 | AK090189.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348245 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146591 | - | - |
| Whole Mouse Genome G4122A | A_51_P430071 | BC047137.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430071 | BC058562.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430071 | AK162494.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430071 | AK167546.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430071 | AK090189.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146591 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02752610 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02752639 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02752591 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22348231 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22348232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02351231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348259 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348265 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348262 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348261 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02425497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02425499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02425491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348268 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348233 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02752597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348236 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348256 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10494389 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1447854_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455095_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10494388 | - | - |
| Mouse Genome 430 2.0 Array | 1447854_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1447854_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1455095_at | - | - |
| Mouse Genome 430 2.0 Array | 1455095_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 319190_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 319190_PM_at | - | - |
| Mouse Expression Set 430 | 1455095_PM_at | - | - |
| Mouse Expression Set 430 | 1455095_at | - | - |
| Murine Genome U74v2 Set | 102558_PM_at | - | - |
| Murine Genome U74v2 Set | 102558_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455095_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447854_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447854_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455095_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 298770 | - | - |
| ABI Mouse Genome Survey Microarray | 433248 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3360184 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3360184 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003360184 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1253851 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231066 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231066 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1253851 | - | - |
| mouse-6 v1.1 (Illumina) | scl00319190.1_155-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3360184 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00319190.1_155-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3360184 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1253851 | - | - |

Hist2h2be
Genomic Map