Assay Details
| Gene Symbol: Npsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175678.3 | NP_783609.1 | 6 - 7 | 764 | 71 | |
| GenBank mRNA | AK033957.1 | - | 6 - 7 | 765 | 71 | |
| AK041451.1 | - | 5 - 6 | 663 | 71 | ||
| AY299387.1 | - | 5 - 6 | 700 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npsr1
Genomic Map