Assay Details
| Gene Symbol: Npsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175678.3 | NP_783609.1 | 8 - 9 | 931 | 126 | |
| GenBank mRNA | AK033957.1 | - | 8 - 9 | 932 | 126 | |
| AK041451.1 | - | 7 - 8 | 830 | 126 | ||
| AK041501.1 | - | 2 - 3 | 227 | 126 | ||
| AY299387.1 | - | 7 - 8 | 867 | 126 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npsr1
Genomic Map