Assay Details
| Gene Symbol: Chn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113246.2 | NP_001106717.2 | 8 - 9 | 1018 | 92 | |
| NM_175752.3 | NP_786928.2 | 2 - 3 | 830 | 92 | ||
| GenBank mRNA | AF332069.1 | - | 2 - 3 | 338 | 92 | |
| AF332070.1 | - | 2 - 3 | 338 | 92 | ||
| AK032630.1 | - | 2 - 3 | 788 | 92 | ||
| AK049943.1 | - | 2 - 3 | 774 | 92 | ||
| AK075606.1 | - | 2 - 3 | 792 | 92 | ||
| AK147686.1 | - | 2 - 3 | 774 | 92 | ||
| BC010825.1 | - | 2 - 3 | 805 | 4219273 | 92 | |
| BC024796.1 | - | 2 - 3 | 781 | 5360438 | 92 | |
| BC025023.1 | - | 2 - 3 | 761 | 5364554 | 92 | |
| BC054770.1 | - | 8 - 9 | 770 | 6415204 | 92 | |
More Information
Set Membership: |
|

Chn1
Genomic Map